Order Entry
Canada
ContactUsLinkComponent
SARS-CoV-2 Recombinant Spike Protein, aa16-685 (HEK293-expressed)
SARS-CoV-2 Recombinant Spike Protein, aa16-685 (HEK293-expressed)
  MSPP-100-0594
 :  
undefined
SARS-CoV-2 Recombinant Spike Protein, aa16-685 (HEK293-expressed)
  MSPP-100-0594
 :  100-0594
 :  

 

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    #N/A
  • Protein/peptide name:
    CoV-2 Recombinant Spike Protein, aa16-685
  • Purity:
    ≥85%
  • Cat. no.:
    MSPP-100-0594

 

 

SARS-CoV-2 Recombinant Spike Protein, aa16-685 is expressed in HEK293 cells and is one of four structural proteins encoded by the SARS-CoV-2 genome. The Spike Protein plays a key role in attachment to host cells, allowing invasion through clathrin-mediated endocytosis. The Spike Protein can be cleaved by host cell proteases after aa685 to yield the N-terminal S1 subunit and C-terminal S2 region. The S1 subunit is responsible for interacting with the host cell receptor (angiotensin-converting enzyme II) through a receptor-binding domain that is highly conserved with SARS-CoV. The S1 subunit has two conformations: a ‘down’ conformation in which the receptor is inaccessible, and an ‘up’ conformation in which the receptor is accessible. These conformational changes are key for monoclonal antibody drugs and vaccine development. SARS-CoV-2 Recombinant Spike Protein contains a polyhistidine tag at the amino terminus; it also contains a FLAG tag at the carboxy terminus.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').