Order Entry
Canada
ContactUsLinkComponent
Human Recombinant PDGF-CC
Human Recombinant PDGF-CC
Catalog # MSPP-781681006
CAS Number:  
undefined
Human Recombinant PDGF-CC
Catalog # MSPP-781681006
Supplier Number:  78168.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/peptide name:
    PDGF-CC
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-781681006

Specifications

About this item

The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer (PDGF-AB; Li and Eriksson). PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin such as fibroblasts and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). PDGF-CC is secreted as a latent growth factor and requires activation by proteolytic processing (Li and Eriksson). PDGF-CC binds to PDGFRα homodimers and PDGFRαβ heterodimers, but not to PDGFRβ homodimers (Li and Eriksson). PDGF-CC is an angiogenic factor that stimulates coronary artery smooth muscle cell proliferation and plays a role in cardiovascular development (Gilbertson et al.). PDGF-CC is also expressed in many tumors and plays a role in tumorigenesis (Zwerner and May).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').