Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant FGF-21
Mouse Recombinant FGF-21
Catalog # MSPP-781081006
CAS Number:  
Mouse Recombinant FGF-21
Catalog # MSPP-781081006
Supplier Number:  78108.1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    50 µg
  • Protein/peptide name:
    FGF-21
  • Purity:
    ≥97%
  • Cat. no.:
    MSPP-781081006

Specifications

About this item

Fibroblast growth factor 21 (FGF-21) is a member of the FGF family. Using β-Klotho as a cofactor, FGF-21 signals through FGF receptor 1c and 4 to activate PI3K and MAPK pathways (Mattila and Härkönen; Kharitonenkov et al.). FGF-21 expression is regulated by tissue-specific peroxisome proliferator-activated receptors (PPARs). Upon PPAR-α stimulation FGF-21 is produced in the liver, and activation of PPAR-γ leads to FGF-21 production in adipose tissue. FGF-21 promotes insulin-independent glucose uptake and lipid accumulation in primary human adipocytes and in mouse 3T3-L1 cells. In pancreatic islets and INS-1 cells it inhibits glucose-mediated glucagon release and stimulates insulin production. FGF-21 does not induce proliferation in immortalized cell lines, unlike other FGFs (Kharitonenkov and Shanafelt). FGF-21 regulates thermogenesis in white and brown adipose tissue, and metabolic processes in cells of pancreatic origin (Kharitonenkov et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').