Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-6R alpha
Human Recombinant IL-6R alpha
Catalog # MSPP-780831006
CAS Number:  
Human Recombinant IL-6R alpha
Catalog # MSPP-780831006
Supplier Number:  78083.1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/peptide name:
    IL-6R alpha
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780831006

Specifications

About this item

Interleukin 6 receptor (IL-6R) alpha is a type I transmembrane glycoprotein that forms a complex with type I transmembrane signal transducer protein gp130 (CD130) and mediates the biological activities of IL-6. IL-6 binds to the membrane-bound non-signaling IL-6R alpha (mIL-6R), and the complex binds to two molecules of gp130 and leads to ‘classical’ IL-6-signal transduction, which includes activation of JAK/STAT, ERK, and PI3K signal transduction pathways (Scheller et al.). In contrast, a soluble form of IL-6R alpha (sIL-6R), which comprises the extracellular portion of the receptor, binds to the secreted IL-6 to form a complex that promotes bioavailability of IL-6. The complex of IL-6 and sIL-6R can bind to gp130 on cells that do not express the IL-6R and are unresponsive to IL-6. This process is known as trans-signaling (Hunter and Jones; Rose-John S). sIL-6R regulates both local and systemic IL-6-mediated events. Elevated levels of sIL-6R have been documented in several disease conditions such as rheumatoid arthritis, myeloma, and Crohn’s disease (Jones et al.; Mihara et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').