If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:1 Unit
- Protein/peptide name:HHV1 (gD) Peptide Pool
- Purity:Average 70%
- Cat. no.:MSPP-100-1406
Stimulating T cells with HHV1 (gD) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV1 (gD) Peptide Pool is a lyophilized mixture of 96 peptides from envelope glycoprotein D of human herpesvirus 1 (HHV1; strain Patton), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 394 on gD. gD is required at the first stage of virus-induced membrane fusion (Subramanian and Geraghty), which likely occurs via binding to receptors such as nectin-1 (Giovine et al.) and HVEM (Connolly et al.) on cell surfaces. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.