If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:1 Unit
- Protein/peptide name:EBV (GP350/GP340) Peptide Pool
- Purity:Average 70%
- Cat. no.:MSPP-100-1390
Stimulating T cells with EBV (GP350/340) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (GP350/340) Peptide Pool is a lyophilized mixture of 224 peptides from envelope glycoprotein GP350 (GP350/340) of Epstein-Barr virus (EBV; strain B95-8), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 907 on GP350/340. GP350 meditates EBV attachment to B lymphocytes via binding to the cell surface receptor CR2 (also known as C3d receptor or CD21) (Nemerow et al.; Tanner et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.