If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:1 unit
- Protein/peptide name:EBV (EBNA-3A) Peptide Pool
- Purity:Average 70%
- Cat. no.:MSPP-100-1386
Stimulating T cells with EBV (EBNA-3A) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (EBNA-3A) Peptide Pool is a lyophilized mixture of 234 peptides from Epstein-Barr nuclear antigen 3 (EBNA-3A) of Epstein-Barr virus (EBV; strain B95-8) and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 944 on EBNA-3A. EBNA-3A contributes to B cell growth transformation (Hertle et al.; Tomkinson et al.) and has been found to interact with host cell proteins, where it activates (Young et al.) or represses (Cludts and Farrell; Hickabottom et al.) transcription of specific cellular genes. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.