Order Entry
Canada
ContactUsLinkComponent
HIV (HLA Class I Control) Peptide Pool
HIV (HLA Class I Control) Peptide Pool
Catalog # MSPP-100-1384
CAS Number:  
undefined
HIV (HLA Class I Control) Peptide Pool
Catalog # MSPP-100-1384
Supplier Number:  100-1384
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 Unit
  • Protein/peptide name:
    HIV (HLA Class I Control) Peptide Pool
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-100-1384

Specifications

About this item

Stimulating T cells with HIV (HLA Class I Control) Peptide Pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HIV Peptide Pool is a lyophilized mixture of 22 peptides from human immunodeficiency virus (HIV), and consists of defined HLA class I-restricted T cell epitopes. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').