Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Hepassocin, His tag
Human Recombinant Hepassocin, His tag
Catalog # MSPP-100-1298
CAS Number:  
undefined
Human Recombinant Hepassocin, His tag
Catalog # MSPP-100-1298
Supplier Number:  100-1298
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    FGL1|HFREP1|HP-041|LFIRE1|LFIRE-1
  • Protein/peptide name:
    Hepassocin
  • Purity:
    ≥87%
  • Cat. no.:
    MSPP-100-1298

Specifications

About this item

Use hepassocin to bind to lymphocyte-activation gene 3 (LAG-3) in an MHC class II independent manner, and inhibit antigen-specific T-cell activation (Wang et al.). Hepassocin is known to play a restorative role in the liver, reducing apoptosis and accelerating hepatocyte proliferation in vivo (Li et al.). In addition to these hepatoprotective effects, studies have shown that hepassocin expression is upregulated in gastric cancer tissues (Zhang et al.) and in breast cancer cells (Du et al.), suggesting it has potential to predict cancer disease progression. Hepassocin is a member of the fibrinogen superfamily, whose members share a fibrinogen domain at their C-terminus. It is predominantly expressed in the liver, and weakly in the pancreas (Hara et al.), and is secreted as a homodimer that consists of 312 amino acids. Hepassocin is an acute-phase reactant whose expression in HepG2 cells has been shown to be regulated by IL-6 (Liu and Ukomadu). This protein product contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, hepassocin from STEMCELL comes lyophilized with ≥ 87% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').