About this item
Interleukin 15 (IL-15) is a four-alpha helix bundle cytokine with many similar properties to IL-2. The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra (the high-affinity receptor for IL-15), as well as IL-2/15Rb (CD122) and common gamma chain (CD132). IL-15 binds to IL-15Rα receptor and can then be presented in trans to IL-2/15Rb and common gamma chain on other cells. Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans (Castillo et al.). The cytoplasmic domains of IL-2/15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways. IL-15 supports the survival and proliferation of naïve CD4+ and CD8+ T cells, and promotes homeostasis of memory T cells. IL-15 also promotes the survival and differentiation of NK cells and regulates their cytolytic activity (Ma et al.). This product is animal component-free.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:IL-15, ACF
- Purity:≥95%