Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor (IL-7R) composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and regulatory T cells (Carrette and Surh). Although a deficiency in IL-7R still permits the generation of normal numbers of peripheral B cells in humans, stimulation of human B cell precursors with IL-7 could promote STAT5-dependent proliferation and survival in vitro (Clark et al.; Corfe and Paige). This product is animal component-free.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Interleukin-7|LP-1|Lymphopoietin 1|Pre-B cell factor
- Protein/peptide name:IL-7, ACF
- Purity:≥95%