Order Entry
Canada
ContactUsLinkComponent
Human Recombinant TNF-alpha, ACF
Catalogue # MSPP-781571006
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
Human Recombinant TNF-alpha, ACF
Catalogue # MSPP-781571006
Fournisseur:  STEMCELL Technologies
Supplier Number:  78157.1
Numéro CAS:  

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein/peptide name:
    TNF-alpha, ACF
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-781571006

Spécifications

À propos de cet article

Tumor necrosis factor-α (TNF-α) is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts. TNF-α has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immuno- suppressive responses. TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová and Hosek). Other effects of TNF-α include vasodilatation and edema formation. This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').