Flt3/Flk-2 (Fms-like tyrosine kinase 3/fetal liver kinase-2) Ligand is a hematopoietic cytokine that plays an important role as a co-stimulatory factor in the proliferation, differentiation, and survival of hematopoietic stem and progenitor cells and the development of the immune system (Hannum et al.). Flt3/Flk-2 Ligand, together with stem cell factor and thrombopoietin, is commonly used to promote expansion of primitive CD34+ hematopoietic cells in culture. In combination with myeloid cytokines such as GM-CSF, G-CSF, or M-CSF, Flt3/Flk-2 Ligand enhances the growth and numbers of clonogenic myeloid progenitor cells. In synergy with the interleukins IL-3, IL-4, IL-7, IL-11, IL-12, IL-15, and GM-CSF and TNF-α, Flt3/Flk-2 Ligand regulates the development of various lymphoid progenitor cells, including dendritic cell, B cell, T cell, and NK cell progenitors. In contrast, Flt3/Flk-2 Ligand has no significant effect on erythropoiesis or megakaryopoiesis (Drexler and Quentmeier; Wodnar-Filipowicz). Flt3/Flk-2 Ligand exists as membrane-bound and soluble isoforms. Both isoforms are biologically active and signal through the class III tyrosine kinase receptor (Flt3/Flk-2, CD135; Rosnet et al.). Flt3/Flk-2 Ligand is produced by a variety of cell types, including uncommitted and committed hematopoietic cells and stromal fibroblasts, whereas expression of the Flt3/Flk-2 receptor is restricted to CD34+ hematopoietic stem and progenitor cells. Flt3/Flk-2 receptor is also expressed on leukemic cells and outside the hematopoietic system in the brain, placenta, and testis (Drexler and Quentmeier; Hannum et al.). This product is animal component-free.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:Flt3/Flk-2 Ligand, ACF
- Purity:≥95%