To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Protein/peptide type:Recombinant
- Species:Mouse
- Size:20 µg
- Protein/peptide name:IL-2
- Purity:≥95%
- Cat. no.:MSPP-78081
Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor (Gaffen and Liu). It binds to a heterotrimeric receptor consisting of CD25, CD122, and CD132. Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.). The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts. IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ regulatory T cells, and affects development of follicular helper T cells. IL-2 also controls inflammation by inhibiting Th17 differentiation (Banchereau et al.). Targeted deletions of the IL-2 gene in mice resulted in development of autoimmune hemolytic anemia, followed by ulcerative colitis. Similar effects were observed in mice that were deficient in IL-2 receptor α (Gaffen and Liu).