Order Entry
Canada
ContactUsLinkComponent
Rat Recombinant GM-CSF
Rat Recombinant GM-CSF
Catalog # MSPP-780181006
CAS Number:  
Rat Recombinant GM-CSF
Catalog # MSPP-780181006
Supplier Number:  78018.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Size:
    20 µg
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780181006

Specifications

About this item

Granulocyte-macrophage colony-stimulating factor (GM-CSF) promotes the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages. In synergy with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, it also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.). GM-CSF is produced by multiple cell types, including stromal cells, Paneth cells, macrophages, dendritic cells (DCs), endothelial cells, smooth muscle cells, fibroblasts, chondrocytes, and Th1 and Th17 cells T cells (Francisco-Cruz et al.). The receptor for GM-CSF (GM-CSFR) is composed of two subunits: the cytokine-specific α subunit (GMRα; CD116) and the common subunit βc (CD131) shared with IL-3 and IL-5 receptors (Broughton et al.). GM-CSFR is expressed on hematopoietic cells, including progenitor cells and immune cells, as well as non-hematopoietic cells. GM-CSF is able to stimulate the development of DCs that ingest, process, and present antigens to the immune system (Francisco-Cruz et al.). Recombinant rat GM-CSF is reactive with mouse cells (Oaks et al.; Vandenabeele et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').