Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/peptide type:Recombinant
- Species:Human
- Size:1000 µg
- Protein synonyms:Basic fibroblast growth factor|FGF-β|FGF2|Fibroblast growth factor-basic|HBGF-2
- Protein/peptide name:bFGF
- Purity:95%
- Cat. no.:MSPP-780031997
Basic fibroblast growth factor (bFGF) is a prototypic member of the fibroblast growth factor family. Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman and Klagsbrun; Kimelman and Kirschner) and are involved in a variety of biological processes including cell proliferation, differentiation, survival, and apoptosis (Folkman and Klagsbrun; Klagsbrun; Rifkin and Moscatelli). bFGF has the β-trefoil structure (Ponting and Russell), binds to the four FGF receptor (FGFR) family members, and activates JAK/STAT, PI3K, ERK1/2, and other receptor tyrosine kinase (RTK) signaling pathways. It supports the maintenance of undifferentiated human embryonic stem cells (Xu et al.; Kang et al.), stimulates human embryonic stem cells to form neural rosettes (Zhang et al.), and improves proliferation of human mesenchymal stem cells and enhances chondrogenic differentiation (Solchaga et al.).