About this item
Interferon-gamma (IFN-γ), also known as type II interferon, is produced by T and NK cells, and in smaller amounts by dendritic cells and macrophages. IFN-γ is controlled by cytokines such as IL-12 and IL-18 secreted in response to infection (Schroder et al.). IFN-γ binds to a receptor complex and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control a diverse array of immunological functions (de Weerd and Nguyen; Krause et al.). IFN-γ stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils (Billiau and Matthys) by promoting the activation of microbial effector functions such as production of reactive oxygen species, NO intermediates, and complement (Schroder et al.). IFN-γ enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes, as well as the production of IL-12 by dendritic cells. In B cells, IFN-γ stimulates survival and growth in both mouse and human cells, and redirects B cells from proliferation towards differentiation. IFN-γ favors the development of Th1 vs Th2 cells and stimulates monocyte differentiation and function (Schroder et al.). This product is animal component-free.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:#N/A
- Protein/peptide name:IFN-gamma, ACF
- Purity:≥95%