Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant IHH
  MSPP-781971006
 :  
Human Recombinant IHH
  MSPP-781971006
 :  78197.1
 :  

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/peptide name:
    IHH
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-781971006

 

 

Indian hedgehog (IHH) is one of three proteins in the hedgehog family which includes sonic hedgehog and desert hedgehog. Hedgehog proteins are important signaling molecules, acting through patched and smoothened receptors. During embryonic development, they are expressed in the fetal lung, gut, stomach, liver, kidney, and pancreas. IHH is strongly expressed in cartilage and regions of developing bone, and its signaling regulates growth and differentiation of bone tissue (Marigo et al.; Valentini et al.; St-Jacques et al.). IHH is also involved in yolk sac vasculogenesis, indicating a role in the differentiation of epiblast cells into endothelial and red blood cells (Byrd et al.). IHH induces the expression of parathyroid hormone-related peptide (PTHrP), which negatively regulates IHH during chondrocyte differentiation and proliferation (Vortkamp et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').