Order Entry
Canada
ContactUsLinkComponent
Human Recombinant RANTES (CCL5)
Human Recombinant RANTES (CCL5)
Catalog # MSPP-780991006
CAS Number:  
Human Recombinant RANTES (CCL5)
Catalog # MSPP-780991006
Supplier Number:  78099.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein/peptide name:
    RANTES (CCL5)
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780991006

Specifications

About this item

RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation (Schall et al.). RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells (Aldinucci and Colombatti; Soria and Ben-Baruch). This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5. RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils. In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic (Lv et al.; Maghazachi et al.). It has been shown that RANTES produced by CD8+ T cells inhibits HIV infection of primary human peripheral blood mononuclear cells (Appay and Rowland-Jones; Cocchi et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').