Order Entry
Canada
ContactUsLinkComponent
Recombinant B18R Protein
Recombinant B18R Protein
Catalog # MSPP-780751006
CAS Number:  
Recombinant B18R Protein
Catalog # MSPP-780751006
Supplier Number:  78075.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    500 µg
  • Protein/peptide name:
    B18R Protein
  • Purity:
    ≥98%
  • Cat. no.:
    MSPP-780751006

Specifications

About this item

The B18R protein, which is encoded by the B18R open reading frame in the Western Reserve (WR) strain of vaccinia virus, is a type I interferon (IFN)-binding protein that is known to inhibit IFN antiviral response (Colamonici et al.). B18R protein has been shown to exhibit neutralizing activity to type I interferon family members (Meyers et al.) and to protect cells from the antiviral effects of interferon (Vancová et al.). It was recently shown that HIV-1–dependent induction of type I IFN activity and BAFF protein levels was abrogated by the soluble B18R protein, with no effect observed on the cell viability (Gomez et al.). B18R protein has been reported to aid in cellular reprogramming using RNA-mediated gene delivery (Warren et al.; Yoshioka N et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').