Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IL-17A
Mouse Recombinant IL-17A
Catalog # MSPP-780331006
CAS Number:  
undefined
Mouse Recombinant IL-17A
Catalog # MSPP-780331006
Supplier Number:  78033.1
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    25 µg
  • Protein/peptide name:
    IL-17A
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780331006

Specifications

About this item

Interleukin 17A (IL-17A) is the founding member of the family of cytokines that includes Interleukin 17B through Interleukin 17F. It is a potent proinflammatory cytokine that plays a key role in defense against pathogens. IL-17A and IL-17F signal as homodimers or heterodimers through the same receptor, and activate NF-kB, MAPK, and C/EBP pathways (Gaffen). IL-17A receptor is expressed on a variety of cell types, including hematopoietic cell compartments. IL-17A is produced by T helper 17 cells, CD8+ T cells, γδ T cells, natural killer T cells, B cells, neutrophils, innate lymphoid cells and mesenchymal stromal cells (MSCs; Zenobia and amp; Hajishengallis; Mojsilovic et al.). IL-17A receptor is expressed at particularly high levels on stromal cells, including MSCs. IL-17A increases the frequency and the average size of colony-forming units-fibroblast derived from bone marrow, as well as the proliferation of bone marrow-derived MSCs. IL-17A suppresses osteogenic differentiation and bone formation of bone marrow-derived MSCs. The action of IL-17A on hematopoiesis is deeply reliant on the microenvironment and the induction of other regulators. In healthy mouse bone marrow, IL-17A stimulates myeloid and early stage erythroid progenitor cells but inhibits late stage erythroid progenitor cells (Mojsilovic et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').