Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IL-13
Mouse Recombinant IL-13
Catalog # MSPP-780301006
CAS Number:  
Mouse Recombinant IL-13
Catalog # MSPP-780301006
Supplier Number:  78030.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    10 µg
  • Protein/peptide name:
    IL-13
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780301006

Specifications

About this item

Interleukin 13 (IL-13) is a type I cytokine, and signals through type I cytokine receptors to activate JAK/STAT and IRS-1/IRS-2 pathways. IL-13 is produced by T cells and innate lymphoid cells (Pulendran and Artis) and it inhibits production of pro-inflammatory cytokines, prostaglandins, and reactive oxygen and nitrogen species by monocytes and macrophages (Hershey). Unlike human B cells, IL-13 has no effect on mouse B cell development and function. IL-13 promotes eosinophil survival, activation, and recruitment, and also activates mast cells (Hershey). IL-13 regulates gastrointestinal parasite expulsion, airway hyperresponsiveness, allergic inflammation, tissue eosinophilia, goblet cell hyperplasia, intracellular parasitism, tissue remodeling, tumor cell growth, and fibrosis. IL-13 induces hypercontractility of smooth muscles by directly acting on the muscle cells as well as the enteric nerves (Wynn).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').