Order Entry
Canada
ContactUsLinkComponent
Human Recombinant VEGF-C
Human Recombinant VEGF-C
Catalogue # MSPP-782021006
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
Human Recombinant VEGF-C
Catalogue # MSPP-782021006
Fournisseur:  STEMCELL Technologies
Supplier Number:  78202.1
Numéro CAS:  

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein synonyms:
    Flt4-L|Flt4 ligand|Vascular endothelial growth factor C|Vascular endothelial growth factor-related protein|VRP
  • Protein/peptide name:
    VEGF-C
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-782021006

Spécifications

À propos de cet article

Vascular endothelial growth factor C (VEGF-C) is a member of the VEGF/platelet-derived growth factor (PDGF) family of proteins. VEGF-C is a potent angiogenic factor and promotes lymphangiogenesis, endothelial cell growth and survival, and can affect blood vessel permeability. VEGF-C is expressed in a range of tissues, but is not expressed in peripheral blood lymphocytes. VEGF-C forms a non-covalent, cell surface-associated, disulfide-linked homodimer that can bind and activate VEGF receptors 2 (VEGFR-2 [Flk1]) and 3 (VEGFR-3 [Flt4]). Interaction with VEGFR-2 results in physiological and intratumoral neoangiogenesis and vessel sprouting (Cao et al.; Tammela et al.), whereas interaction with VEGFR-3 is critical for lymphangiogenesis (Karkkainen et al.; Laakkonen et al.; Mäkinen et al.). Overexpression of VEGF-C in tumor cells has been shown to result in enhanced lymph flow and increased metastasis to regional lymph nodes (Hoshida et al.; Mandriota et al.; Padera et al.; Skobe et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').