Order Entry
Canada
ContactUsLinkComponent
Human Recombinant MIP-4
Human Recombinant MIP-4
Catalog # MSPP-100-1334
CAS Number:  
undefined
Human Recombinant MIP-4
Catalog # MSPP-100-1334
Supplier Number:  100-1334
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    #N/A
  • Protein/peptide name:
    MIP-4
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-100-1334

Specifications

About this item

Macrophage inflammatory protein-4 (MIP-4) plays a role in adaptive and innate immune responses, acting as a chemoattractant for T and B lymphocytes, and natural killer cells (Krohn et al.). MIP-4 can also induce immune tolerance through its effects on dendritic cells and macrophages (Azzaoui et al., Schraufstatter et al.). In breast cancer, MIP-4 has been shown to promote the invasiveness and metastasis of cancer cells in vivo by binding PITPNM3 (Chen et al.).This small cytokine belongs to the chemokine family found only in primates. Based on amino acid sequence, MIP-4 is closely related to MIP-1 alpha (CCL3) and MIP-1 beta (CCL4) (Liang et al.). MIP-4 is produced by antigen-presenting immune cells including dendritic cells, macrophages, and monocytes (Lieshout et al.). For consistency and reproducibility across your applications, macrophage inflammatory protein-4 from STEMCELL comes lyophilized with ≥95% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').