Order Entry
Canada
ContactUsLinkComponent
Human Recombinant BTN1A1, His Tag
Human Recombinant BTN1A1, His Tag
Catalog # MSPP-100-1327
CAS Number:  
undefined
Human Recombinant BTN1A1, His Tag
Catalog # MSPP-100-1327
Supplier Number:  100-1327
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein synonyms:
    #N/A
  • Protein/peptide name:
    BTN1A1
  • Purity:
    ≥92%
  • Cat. no.:
    MSPP-100-1327

Specifications

About this item

Butyrophilin 1A1 (BTN1A1) is a glycoprotein belonging to the butyrophilin (BTN) family which collectively modulates immune responses, through excitatory and inhibitory signals targeting immune cells. Butyrophilins are composed of two extracellular immunoglobulin domains and a transmembrane region, with a conserved B30.2 domain (PRYSPRY) that is present in most members (Malinowska et al.). BTN1A1, along with BTN2A2, has been shown to inhibit the proliferation of CD4+ and CD8+ T cells, modify T cell metabolism, and affect the expression of IL-2 and IFN-γ (Smith et al.). Due to these modulatory effects on T cells, BTN1A1 may have a role in inhibiting the development of autoimmune diseases (Stefferl et al.). BTN1A1 is also expressed in mammary glands and is required for the secretion of milk lipids during lactation (Ogg et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, Serpin A12 from STEMCELL comes lyophilized with ≥92% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').