Order Entry
Canada
ContactUsLinkComponent
SARS-CoV-2 (Spike Protein) Peptide Pool
SARS-CoV-2 (Spike Protein) Peptide Pool
Catalog # MSPP-100-0676
CAS Number:  
undefined
SARS-CoV-2 (Spike Protein) Peptide Pool
Catalog # MSPP-100-0676
Supplier Number:  100-0676
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 units
  • Protein/peptide name:
    SARS-CoV-2 (Spike Protein) Peptide Pool
  • Purity:
    Average 70%
  • Cat. no.:
    MSPP-100-0676

Specifications

About this item

The SARS-CoV-2 (spike protein) peptide pool is provided as two lyophilized mixtures (subpools) from the SARS-CoV-2 spike protein. Each subpool contains 158 peptides, for a total of 316 peptides. The virus attaches to the cell membrane of the host through the interaction between spike protein and angiotensin-converting enzyme 2 (ACE2) receptor, and the spike protein plays a critical role in viral entry. The subpools consist of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1273 on the spike protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').