If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:1 unit
- Protein/peptide name:RSV (HLA Class I Control) Peptide Pool
- Purity:≥95%
- Cat. no.:MSPP-100-0674
The RSV (HLA Class I Control) peptide pool is a lyophilized mixture of 28 peptides from respiratory syncytial virus (RSV). The pool consists of defined HLA class I-restricted T cell epitopes, and can be used as an antigen-specific positive control to stimulate T cells.