ContactUsLinkComponent
Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Catalog # 60004-228
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Catalog # 60004-228
Supplier:  Kemtech America
Supplier Number:  H200001
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Synthware Heat Transfer Block for Minivials, Microscale
  • Cat. no.:
    60004-228
  • Supplier No.:
    H200001
Specifications
About this item
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
  • Aluminum heat transfer block
  • Hold three sizes of vials
Block has two holes for 3 ml and 5 ml heavy wall reaction vials, four holes for Craig recrystallization tubes, and one hole for a standard thermometer.