Order Entry
Canada
Orders LinkContactUsLinkComponent
RNA Marker

RNA Marker

RNA marker, 0.5 - 9 kb, 50 µg, 1 mg/ml
Catalog # CA12001-748
Supplier:   Lonza
Supplier #:  50575
RNA Marker
Catalog # CA12001-748
Supplier:  Lonza
CAS Number:  
RNA Marker
RNA Marker

RNA marker, 0.5 - 9 kb, 50 µg, 1 mg/ml

CAS Number:  
Price
$723.94
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

RNA standards may be denatured with standard procedures, and can be visualized in Northern blots with labeled lambda sequence.

  • Ethidium bromide staining will easily detect 4 µg on a standard agarose gel
  • Smaller quantities can be detected with GelStar® Stain or SYBR® Green II Gel Stain

These RNA standards may be denatured with standard procedures, and can be visualized on Northern blots with labeled lambda sequence. Markers are provided at a concentration of 1 mg/ml. Ethidium bromide staining will easily detect 4 µg (4 µl) on a standard agarose gel. Smaller quantities can be detected with GelStar® nucleic acid gel stain or SYBR® green II gel stain.

RNA markers consist of RNA transcripts from 0.5 to 9 kb, in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9 kb.

Specifications

Specifications

  • Concentration:
    1 mg/ml
  • Description:
    RNA marker, 0.5 - 9 kb, 50 µg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.