BioRobotix® Pipette Tips for MultiMek™ Workstations, Molecular BioProducts
Supplier: Thermo Scientific
Certificates
About this item
Compatible with MultiMek™ style workstations/AP96.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as clear polypropylene only, in 20, 130 and 250 µl sizes. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Specifications
- Catalog No:
- 72830-224
- 72830-220
- 72830-230
- 72830-228
- 72830-226
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)