UriScan Urine Test Strips, BioSys
Supplier: BioSys
Certificates
About this item
Test strips for urinalysis testing of various parameters
- 100 strips per vial
Strips are compatible with the BioSys UriScan Optima II and UriScan Pro Urine analyzers as well as Bayer® and Roche® analyzers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.