Model 8100 Variable Repetitive Syringe Dispenser, Nichiryo America
Catalog #: 89078-434
Supplier: Nichiryo America
Specifications
- Volume:1 - 5000 µl
- Description:Syringe Dispenser
- Cat. no.:89078-434
- Supplier No.:8100
Specifications
About this item
The lightweight and slender Nichiryo 8100 syringe dispenser provides 35 methods of liquid dispensing throughout the 1 to 5000 µl volume range.
The ergonomic design permits easy handling without fatigue. Syringes are quickly and easily interchangeable, and volume setting is conveniently adjustable by dial. Dispensers offer excellent accuracy and reproducibility. With the exception of the 3 and 60 ml units, all syringes are autoclavable. Syringe 89078-442 includes a spacer in the nozzle specially designed for use with low liquid pressure microtiter plates. Syringe 89078-450 includes one adaptor to mount syringe to dispenser handle.
Customers Who Bought This Also Bought