Axygen® Pipette Tip Reload System, Corning
Supplier: Corning
Certificates
About this item
Gilson®-style and universal fit pipette tips in easy-to-load refill racks.
- RNase-/DNase-free
- Nonpyrogenic
- Translucent rack allows tips to be easily seen
- Color-coded insert allows for easy identification of tip
Universal fit tips have a beveled reference and graduated marks.
Axygen® tip reload system contains durable refill rack that can be easily refilled by hand and is autoclavable. They are available in different colors and are packaged in bulk or racks.