Pure™ 1000 SoftFit™ Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
Certificates
About this item
The SoftFit design of these tips ensures tip-to-tip precision and universal fit.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Nonsterile tips are autoclavable
This feature also makes loading and ejecting easy and minimizes hand fatigue. Tips are compatible with most major brand pipettors.
Ordering information: Tips 53225-712 and -710 are packaged 100 per tray, 8 trays per pack. Tips 53225-708 are bulk packaged.
Specifications
- Catalog No:
- 53225-712
- 53225-710
- 53225-708
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
Specifications
Related Information
Guide to ART Pipet Tips
Videos
Thermo Scientific ART Universal Tips
Product Literature
Frequently Bought Together