Pure™ XLG Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
Certificates
About this item
The Pure XLG tips are ideal for larger, more viscous samples.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
- Non-sterile tips are autoclavable
Features include extended tip length and large orifice. Designed specifically for the pipetting of fragile cells, viscous samples, and genomic DNA. Tips are compatible with Pipetman*, Eppendorf*, Socorex*, and Rainin* pipettors.
Ordering information: Tips are packaged 96 per tray, 8 trays per pack.