Order Entry
Canada
ContactUsLinkComponent
Ultrasensitive Insulin ELISA
Ultrasensitive Insulin ELISA
Catalog # 102977-824
Supplier:  Alpco Diagnostics
CAS Number:  
Ultrasensitive Insulin ELISA
Catalog # 102977-824
Supplier:  Alpco Diagnostics
Supplier Number:  80INSHUUE01.1
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Insulin
  • Description:
    Ultrasensitive insulin ELISA
  • Size:
    1×96 tests
  • Sample type:
    Plasma, serum
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h, 30 min
  • Detection range:
    0.15 - 20 µIU/ml
  • Sensitivity:
    0.135 µIU/ml
  • Regulatory status:
    FDA Class I Device. For in vitro diagnostic use.
  • Cat. no.:
    102977-824
  • Supplier No.:
    80INSHUUE01.1

Specifications

About this item

The Ultrasensitive Insulin ELISA quantifies the concentration of insulin protein in human serum or plasma samples.

  • Only 25 µl of sample is sufficient for the assay
  • Superior sensitivity of 0.135 µIU/ml
  • Dynamic range of 0.15 to 20 µIU/ml

The Ultrasensitive Insulin ELISA kit is a FDA registered in vitro diagnostic tool for the quantification of human insulin in a clinical setting or research laboratory. The Ultrasensitive Insulin ELISA utilizes a dual-monoclonal antibody sandwich ELISA format, diabetes control, and 96-well microplate comprising removable strips. The Ultrasensitive Insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if the entire kit is not needed at one time.

Certifications: FDA Class I Device.
Packaging: This kit is also available in a jumbo (10-plate) format.
Caution: For In Vitro diagnostic use.