C-peptide ELISA Jumbo (10-Plate)
102977-820
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:C-peptide
- Description:C-peptide ELISA Jumbo (10-plate)
- Size:10×96 tests
- Environmentally Preferable:
- Sample type:Plasma, serum
- Detection method:Colorimetric
- Time to Results:2 h
- Detection range:20 - 3000 pmol/L
- Sensitivity:2.95 pmol/L
- Regulatory status:FDA Class I Device. For in vitro diagnostic use.
- Cat. no.:102977-820
- Supplier No.:80-CPTHU-E10
The C-peptide ELISA tool for the quantification of human C-peptide in serum and plasma samples in both clinical and research laboratory settings.
: FDA Class I Device.
: For In Vitro diagnostic use.