Biohit Family Extended Length Pipette Tips, Sartorius
The non-filtered Optifit tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision.
- Tips work with most brands of pipettors
- Autoclavable at 121 °C (250 °F) for 20 minutes
- Single-handed opening and closing of flip-top racks
- E-beam presterilized for sterile work
Use to reach solutions in flasks, deep well plates, tubes, and other difficult vessels
These pipette tips guarantee accurate results when used with Biohit® pipettors.
The tips work with most popular brands of single and multichannel pipettors. They also fit most research-grade pipettors.
Frequently Bought Together