BioWhittaker® Hanks’ Balanced Salt Solutions, Lonza
Supplier: Avantor
Certificates
About this item
All balanced salt solutions contain sodium bicarbonate and are 1X
Store at 15 to 30°C (59 to 86°F). Manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility.
Ordering information: Solution 12002-008 is provided in a flexible package with one female luer connector, one blood spike receptacle, and one male MPC connector with cap.