To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pierce Erase-It® Background Eliminator Kit, Sufficient Reagent to Prepare 3 L of Working Solution
- Environmentally Preferable:
- Cat. no.:CAPI21065
- Supplier No.:21065
Specifications
About this item
The background eliminator reduces background signal evenly over any film within minutes
- Fast, easy background elimination from film
- Eliminates unwanted background from overexposed, speckled or shaded films
- No need for time-consuming re-exposures to find the optimal image
- No need to re-optimise assay reagents in order to obtain the optimal image
The eliminator will work with any application that requires X-ray film exposures, and it is compatible with both newly developed films or films that have been stored for years. Simply immerse the exposed film in the working solution, watch for the desired image, and stop the reaction by rinsing the film with water. There is no need to re-expose the blot or even completely re-run the experiment.
High background, shading, overexposed bands and speckling are problems inherent to film exposure. The Erase-It® kit can correct these problems without the need to re-expose the blot to film or re-do the experiment. Erase-It® solution can be used with newly exposed films or exposed films that have been stored. It can also be used with any brand of film.