Acrodisc™ Syringe Filters with PVDF Membrane, Cytiva (Formerly Pall Lab)
: Cytiva
Acrodisc™ PVDF syringe filters, can be used for both aqueous and non-aggressive solvent-based sample filtration applications.
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
- Reduces experimental variables
- HPLC certified for low levels of UV-absorbing extractables
- Protects columns and instrumentation against particulate build up
The Acrodisc™ syringe filter with a 13 mm minispike configuration offers low hold-up and easy filtration into autosampler vials.
- Catalog No:
- CA77553-304
- CA77553-302
- CA77553-282
- CA77553-280
- CA77553-300
- CA77553-306
- CA77553-274
- Type:
- CA77553-304
- CA77553-302
- CA77553-282
- CA77553-280
- CA77553-300
- CA77553-306
- CA77553-274
- Hold-up volume:
- <14 µl
- <30 µl
- <125 µl
- <100 µl
- <30 µl
- <14 µl
- <125 µl
- Max. operating pressure:
- 50.7 psi
- 50.7 psi
- 30.4 psi
- 30.4 psi
- 50.7 psi
- 50.7 psi
- 59.4 psi
- Max. temperature:
- 55 °C
- 55 °C
- 82 °C
- 82 °C
- 55 °C
- 55 °C
- 82 °C
- Water flow rate:
- 15 ml/min at 3.1 bar (310 kPa, 45 psi)
- 5 ml/min at 3.1 bar (310 kPa, 45 psi)
- 144 ml/min at 2.1 bar (210 kPa, 30 psi)
- 144 ml/min at 2.1 bar (210 kPa, 30 psi)
- 15 ml/min at 3.1 bar (310 kPa, 45 psi)
- 5 ml/min at 3.1 bar (310 kPa, 45 psi)
- 144 ml/min at 2.1 bar (210 kPa, 30 psi)
Frequently Bought Together