Corning® BioCoat® Collagen I German Glass Coverslip, 22 mm, Bulk Packed, Corning
Catalog #: CACB354089
Supplier: Corning
Specifications
- Description:German glass coverslip, round
- Material:Glass
- Cat. no.:CACB354089
- Supplier No.:354089
Specifications
About this item
Corning® BioCoat™ 22 mm #1 German Glass coverslip with a uniform application of rat tail collagen. Bulk packed, 60/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
- Shelf life: >= 3 months from date of shipment
Customers Who Bought This Also Bought