About this item
For applications that require high accuracy and reproducibility.
Manufactured using diamond-polished molds to eliminate the surface cavities and occlusions which cause sample retention. Polymer resists DNA, RNA, and protein adherence and will not denature DNA. Manufactured without secondary substances such as silicone or heavy metals, which can contaminate samples.
- Compatible with most major brands of pipettors
For applications that require high accuracy and reproducibility.
Manufactured using diamond-polished molds to eliminate the surface cavities and occlusions which cause sample retention. Polymer resists DNA, RNA, and protein adherence and will not denature DNA. Manufactured without secondary substances such as silicone or heavy metals, which can contaminate samples.
Manufactured using diamond-polished molds to eliminate the surface cavities and occlusions which cause sample retention. Polymer resists DNA, RNA, and protein adherence and will not denature DNA. Manufactured without secondary substances such as silicone or heavy metals, which can contaminate samples.
Tips are autoclavable and presterilized through exposure to e-beam radiation for protection from contamination.
Ideal for DNA multiplication and handling radioactive, biohazardous, and corrosive samples. Filter prevents cross-contamination from the pipette to subsequent samples.
Related Information
Certificates
Search by Lot Number22234-008_07019295 22234-008_32419295 22234-012_12819482 22234-018_29119294 22234-132_02519091