Specifications
- pH Range:0 - 14
- Body Material:Glass
- Internal Reference:Ag/AgCl
- Junction:Ceramic
- Connector:BNC
- Supplier No.:9102DJWP
Specifications
About this item
This double junction pH electrode offers superior measurement stability, faster response, greater accuracy, longer life, least long-term drift, and more reproducible results.
- With waterproof BNC connector
- Ag/AgCl double junction
- Refillable
Refillable pH electrode with a waterproof BNC connector and 1m (393/8") cable. Accurate and reliable. For use in TRIS buffers and with proteins. Works with any pH meter with a BNC connector. Mercury-free and environmentally safe.