Order Entry
Canada
Orders LinkContactUsLinkComponent
Infecon® Absorbent Strips, Com-Pac

Infecon® Absorbent Strips, Com-Pac

10.2×20.3 cm (4×8")
Catalog # 11217-528
Supplier:   Com-Pac
Supplier #:  INF-1004
Infecon® Absorbent Strips, Com-Pac
Catalog # 11217-528
Supplier:  Com-Pac
Infecon® Absorbent Strips, Com-Pac
Infecon® Absorbent Strips, Com-Pac

10.2×20.3 cm (4×8")

Price
$76.28
Log in for availability
Pack of 100
Product Details & Documents

Designed for use with Infecon shippers and diagnostic packaging, these absorbent strips provide fast and reliable protection against spills.

Strips have an absorbent capacity of 250mL.

Documents

Specifications
  • Dimensions:
    10.2×20.3 cm (4×8")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.