pH Storage Bottles, Narrow Neck, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Stores and protects Orion™ pH electrodes.
- Electrode storage bottles can be used with any 6, 8, or 12 mm pH electrodes
- Electrode bulb guards are designed for use with glass body pH electrodes
Storage bottles protect and condition electrodes when not in use, and bulb guards protect the pH sensing bulb of glass pH electrodes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.