Antibiotics and Antimycotics, BD Diagnostics
Supplier: BD
Certificates
About this item
Ideal for the qualitative susceptibility testing of mycobacteria and related organisms.
- Reliable
- Convenient
- Rapid method
This collection of media products is used for the evaluation of susceptibility of microorganisms to anti-microbial agents.
Packaging: Each test includes 50 susceptibility disks per cartridge.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.