Paint Can Shippers, UN Marked, Qorpak®
Supplier: Berlin Packaging
Certificates
About this item
Specially designed combination package for paint shipments as well as an overpack for containers that have not passed internal pressure tests
- One-gallon size is suitable for liquids or solids
- Pressure capability of 95kPa
Meet UN 4G requirements (labels required).
Includes paint can, absorbent material, outer corrugated carton, sealing ring, plastic bag, tape, and instructions.
Frequently Bought Together