Nalgene® Oak Ridge Centrifuge Tubes, Polysulfone, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Strong plastic tubes with good resistance to corrosive bases.
- Leakproof and autoclavable
- Transparent with a yellow tint
- Maximum RCF is 50000 ×g
- Polypropylene screw closures
For standard and refrigerated centrifugation of biological and chemical samples. When autoclaving, place cap loosely on tube without engaging threads.
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Certifications: Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps include a retained silicone gasket in addition to the tube and screw closure. Nominal volumes correspond to rotor tube sizes; please refer to nominal volumes when ordering tubes to fit a rotor.
Customers Who Bought This Also Bought