Order Entry
Canada
ContactUsLinkComponent
Transwell® Dish Inserts, Polycarbonate, Sterile, Corning
Transwell® Dish Inserts, Polycarbonate, Sterile, Corning
  29442-116
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Transwell® Dish Inserts, Polycarbonate, Sterile, Corning
  29442-116
 :  Corning
 :  3420
Product available on GSA Advantage®
   Low Manufacturing Impact

 

  • Ø:
    75 mm
  • Pore size:
    3.0 µm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Cat. no.:
    29442-116
  • Well Area:
    44 cm2
  • Supplier No.:
    3420

 

 

These tissue culture treated polycarbonate inserts are for use in epithelial and endothelial cell culture, transport, migration/invasion, co-culture, and microbial pathogenesis studies

Inserts provide independent access to both the apical and basolateral sides of the cell monolayer.

 : Inserts are supplied with dishes. Individually wrapped.